7-Chloroimidazo[1,2-a]pyrimidin-5(1H)-one 97%

BLDPBD239005-250MGEA 181 EUR
BLDPBD239005-250MG BLDPBD239005-1G BLDPBD239005-100MG
7-Chloroimidazo[1,2-a]pyrimidin-5(1H)-one 97%
7-Chloroimidazo[1,2-a]pyrimidin-5(1H)-one

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₄ClN₃O
MW: 169,57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14706869
CAS Number: 57473-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR