Ethyl 6-chloropyrazine-2-carboxylate 95%

BLDPBD239065-5GEA 385 EUR
BLDPBD239065-5G BLDPBD239065-250MG BLDPBD239065-1G BLDPBD239065-100MG
Ethyl 6-chloropyrazine-2-carboxylate 95%
Ethyl 6-chloropyrazine-2-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₇ClN₂O₂
MW: 186,59 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16036493
CAS Number: 161611-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR