4,7-Dimethoxy-1-naphthaldehyde 95%

BD239152-100MG BD239152-1G BD239152-250MG
BLDPBD239152-100MGEA 42 EUR
BLDPBD239152-100MG BLDPBD239152-1G BLDPBD239152-250MG
4,7-Dimethoxy-1-naphthaldehyde 95%
4,7-Dimethoxy-1-naphthaldehyde

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₁₂O₃
MW: 216,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865183
CAS Number: 90381-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR