2-(2-Methylpyridin-4-yl)ethanamine 95%

BD239358-250MG BD239358-100MG
BLDPBD239358-250MGEA 650 EUR
BLDPBD239358-250MG BLDPBD239358-100MG
2-(2-Methylpyridin-4-yl)ethanamine 95%
2-(2-Methylpyridin-4-yl)ethanamine
Formula: C₈H₁₂N₂
MW: 136,19 g/mol
MDL Number: MFCD09924494
CAS Number: 625438-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR