Ethyl 5-(4-methoxyphenyl)-1,3,4-oxadiazole-2-carboxylate 98%

BD239409-10G BD239409-5G BD239409-250MG BD239409-1G
BLDPBD239409-10GEA 208 EUR
BLDPBD239409-10G BLDPBD239409-5G BLDPBD239409-250MG BLDPBD239409-1G
Ethyl 5-(4-methoxyphenyl)-1,3,4-oxadiazole-2-carboxylate 98%
Ethyl 5-(4-methoxyphenyl)-1,3,4-oxadiazole-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₁₂N₂O₄
MW: 248,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441302
CAS Number: 99367-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR