tert-Butyl 3-(aminomethyl)piperazine-1-carboxylate 95%

BLDPBD239595-100MGEA 116 EUR
BLDPBD239595-100MG BLDPBD239595-250MG
tert-Butyl 3-(aminomethyl)piperazine-1-carboxylate 95%
tert-Butyl 3-(aminomethyl)piperazine-1-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₀H₂₁N₃O₂
MW: 215,29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22056449
CAS Number: 1376099-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR