Methyl 3-(((tert-butoxycarbonyl)amino)methyl)benzoate 97%

BD239656-100MG BD239656-10G BD239656-1G BD239656-5G BD239656-250MG
BLDPBD239656-100MGEA 9.5 EUR
BLDPBD239656-100MG BLDPBD239656-10G BLDPBD239656-1G BLDPBD239656-5G BLDPBD239656-250MG
Methyl 3-(((tert-butoxycarbonyl)amino)methyl)benzoate 97%
Methyl 3-(((tert-butoxycarbonyl)amino)methyl)benzoate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₁₉NO₄
MW: 265,31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14583094
CAS Number: 180863-55-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR