7-Bromo-6-fluoroisoquinoline 97%

BLDPBD240749-5GEA 1080 EUR
BLDPBD240749-5G BLDPBD240749-1G BLDPBD240749-250MG BLDPBD240749-100MG
7-Bromo-6-fluoroisoquinoline 97%
7-Bromo-6-fluoroisoquinoline

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₅BrFN
MW: 226,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846300
CAS Number: 923022-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR