5-Bromo-1H-pyrrole-2-carbaldehyde 95%

BLDPBD240935-100MGEA 53.6 EUR
BLDPBD240935-100MG BLDPBD240935-250MG BLDPBD240935-1G BLDPBD240935-5G
5-Bromo-1H-pyrrole-2-carbaldehyde 95%
5-Bromo-1H-pyrrole-2-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₄BrNO
MW: 173,99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13178669
CAS Number: 931-34-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR