2-Chloro-3,6-dimethylpyridine 95%

BLDPBD241047-250MGEA 108 EUR
BLDPBD241047-250MG BLDPBD241047-1G BLDPBD241047-100MG
2-Chloro-3,6-dimethylpyridine 95%
2-Chloro-3,6-dimethylpyridine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₈ClN
MW: 141,598 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12400985
CAS Number: 72093-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR