1H-Pyrazolo[3,4-c]pyridin-3(2H)-one 98%

BD241124-250MG BD241124-100MG
BLDPBD241124-250MGEA 47.3 EUR
BLDPBD241124-250MG BLDPBD241124-100MG
1H-Pyrazolo[3,4-c]pyridin-3(2H)-one 98%
1H-Pyrazolo[3,4-c]pyridin-3(2H)-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₅N₃O
MW: 135,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176077
CAS Number: 53975-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR