Ethyl benzo[d]oxazole-6-carboxylate 95+%

BLDPBD241219-250MGEA 31.5 EUR
BLDPBD241219-250MG BLDPBD241219-100MG BLDPBD241219-1G
Ethyl benzo[d]oxazole-6-carboxylate 95+%
Ethyl benzo[d]oxazole-6-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₉NO₃
MW: 191,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16036195
CAS Number: 1355171-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR