Methyl 2-(bromomethyl)-5-methoxybenzoate 98%

BD241558-250MG
BLDPBD241558-250MGEA 26.3 EUR
BLDPBD241558-250MG
Methyl 2-(bromomethyl)-5-methoxybenzoate 98%
Methyl 2-(bromomethyl)-5-methoxybenzoate
Formula: C₁₀H₁₁BrO₃
MW: 259,1 g/mol
MDL Number: MFCD11505947
CAS Number: 788081-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR