5-Bromothieno[2,3-d]pyrimidin-4(3H)-one 95%

BLDPBD241777-250MGEA 64.1 EUR
BLDPBD241777-250MG BLDPBD241777-5G BLDPBD241777-100MG BLDPBD241777-1G
5-Bromothieno[2,3-d]pyrimidin-4(3H)-one 95%
5-Bromothieno[2,3-d]pyrimidin-4(3H)-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₃BrN₂OS
MW: 231,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25542166
CAS Number: 1239460-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR