7-Nitroquinolin-2(1H)-one 97%

BD242236-250MG BD242236-100MG
BLDPBD242236-250MGEA 176 EUR
BLDPBD242236-250MG BLDPBD242236-100MG
7-Nitroquinolin-2(1H)-one 97%
7-Nitroquinolin-2(1H)-one

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₆N₂O₃
MW: 190,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18448289
CAS Number: 75755-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR