1,3-Dimethyl-2-imidazolidinone 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 1,3-Dimethylimidazolidin-2-one

BLDPBD2428-25GEA 7.4 EUR
BLDPBD2428-25G
1,3-Dimethyl-2-imidazolidinone 98%
1,3-Dimethyl-2-imidazolidinone

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₁₀N₂O
MW: 114,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003188
CAS Number: 80-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR