Dimethyl 2-methylterephthalate 98%

BD242936-250MG BD242936-5G BD242936-1G
BLDPBD242936-250MGEA 30.5 EUR
BLDPBD242936-250MG BLDPBD242936-5G BLDPBD242936-1G
Dimethyl 2-methylterephthalate 98%
Dimethyl 2-methylterephthalate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₂O₄
MW: 208,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11502730
CAS Number: 14186-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR