1-(4-Formylphenyl)azetidine-3-carboxylic acid 98%

BLDPBD243156-1GEA 200 EUR
BLDPBD243156-1G BLDPBD243156-250MG BLDPBD243156-100MG
1-(4-Formylphenyl)azetidine-3-carboxylic acid 98%
1-(4-Formylphenyl)azetidine-3-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₁H₁₁NO₃
MW: 205,20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21333096
CAS Number: 1355248-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR