Methyl 4'-formyl-[1,1'-biphenyl]-2-carboxylate 97%

BD243254-250MG BD243254-1G BD243254-5G
BLDPBD243254-250MGEA 32.6 EUR
BLDPBD243254-250MG BLDPBD243254-1G BLDPBD243254-5G
Methyl 4'-formyl-[1,1'-biphenyl]-2-carboxylate 97%
Methyl 4'-formyl-[1,1'-biphenyl]-2-carboxylate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₅H₁₂O₃
MW: 240,25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06801752
CAS Number: 144291-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR