5-Fluoro-3-nitrobenzene-1,2-diamine 95%

BD243837-100MG BD243837-250MG
BLDPBD243837-100MGEA 137 EUR
BLDPBD243837-100MG BLDPBD243837-250MG
5-Fluoro-3-nitrobenzene-1,2-diamine 95%
5-Fluoro-3-nitrobenzene-1,2-diamine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₆FN₃O₂
MW: 171,12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26743427
CAS Number: 170098-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR