3-Methoxyisoxazol-5-amine 95%

BLDPBD243866-100MGEA 175 EUR
BLDPBD243866-100MG
3-Methoxyisoxazol-5-amine 95%
3-Methoxyisoxazol-5-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₄H₆N₂O₂
MW: 114,10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12402090
CAS Number: 35143-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR