4-Bromo-5-methoxy-1H-indole 98%

Supplier: BLD PHARMATECH GMBH

BLDPBD244126-1GEA 383 EUR
BLDPBD244126-1G BLDPBD244126-100MG BLDPBD244126-250MG BLDPBD244126-5G
4-Bromo-5-methoxy-1H-indole 98%
4-Bromo-5-methoxy-1H-indole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₈BrNO
MW: 226,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12973810
CAS Number: 90858-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR