2-(Diphenylphosphino)phenol 98%

BLDPBD244508-1GEA 67.2 EUR
BLDPBD244508-1G BLDPBD244508-25G BLDPBD244508-250MG BLDPBD244508-5G
2-(Diphenylphosphino)phenol 98%
2-(Diphenylphosphino)phenol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₈H₁₅OP
MW: 278,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015846
CAS Number: 60254-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR