(S)-3-((tert-Butoxycarbonyl)amino)-3-(6-methoxypyridin-3-yl)propanoic acid 95+%

BLDPBD244695-100MGEA 267 EUR
BLDPBD244695-100MG BLDPBD244695-250MG BLDPBD244695-1G
(S)-3-((tert-Butoxycarbonyl)amino)-3-(6-methoxypyridin-3-yl)propanoic acid 95+%
(S)-3-((tert-Butoxycarbonyl)amino)-3-(6-methoxypyridin-3-yl)propanoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₄H₂₀N₂O₅
MW: 296,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04118073
CAS Number: 1217755-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR