(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(furan-2-yl)propanoic acid 95%

BD244703-1G BD244703-100MG BD244703-250MG
BLDPBD244703-1GEA 360 EUR
BLDPBD244703-1G BLDPBD244703-100MG BLDPBD244703-250MG
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(furan-2-yl)propanoic acid 95%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(furan-2-yl)propanoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₂H₁₉NO₅
MW: 377,39 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04117844
CAS Number: 1217662-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR