4-Aminotetrahydropyran 97%

Supplier: BLD PHARMATECH GMBH

Synonyms: Tetrahydro-2H-pyran-4-amine

BLDPBD2449-10GEA 33.6 EUR
BLDPBD2449-10G BLDPBD2449-1G BLDPBD2449-5G BLDPBD2449-25G
4-Aminotetrahydropyran 97%
4-Aminotetrahydropyran

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₁₁NO
MW: 101,1469 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02179436
CAS Number: 38041-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR