(S)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one 97%

BD245936-1G BD245936-250MG BD245936-100MG
BLDPBD245936-1GEA 301 EUR
BLDPBD245936-1G BLDPBD245936-250MG BLDPBD245936-100MG
(S)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one 97%
(S)-5-Benzyl-2,2,3-trimethylimidazolidin-4-one

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₃H₁₈N₂O
MW: 218,29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20482089
CAS Number: 132278-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR