tert-Butyl 3-amino-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate 98%

BLDPBD246049-250MGEA 141 EUR
BLDPBD246049-250MG BLDPBD246049-1G BLDPBD246049-100MG
tert-Butyl 3-amino-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate 98%
tert-Butyl 3-amino-6,7-dihydro-1H-pyrazolo[4,3-c]pyridine-5(4H)-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₁H₁₈N₄O₂
MW: 238,28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12964106
CAS Number: 398491-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR