3-Hydrazinylquinoxaline-2(1H)-thione 98+%

BLDPBD246387-1GEA 350 EUR
BLDPBD246387-1G
3-Hydrazinylquinoxaline-2(1H)-thione 98+%
3-Hydrazinylquinoxaline-2(1H)-thione

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formula: C₈H₈N₄S
MW: 192,24 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00457014
CAS Number: 13080-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR