3-Formyl-4-methylbenzoic acid 98%

Supplier: BLD PHARMATECH GMBH

Synonyms: 3-Formyl-4-methylbenzoic Acid

BLDPBD246532-100MGEA 56.7 EUR
BLDPBD246532-100MG BLDPBD246532-250MG BLDPBD246532-1G
3-Formyl-4-methylbenzoic acid 98%
3-Formyl-4-methylbenzoic acid

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₈O₃
MW: 164,158 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20660432
CAS Number: 69526-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR