Potassium (5-bromo-2-fluoropyridin-3-yl)trifluoroborate 95%

BLDPBD246918-1GEA 14.7 EUR
BLDPBD246918-1G
Potassium (5-bromo-2-fluoropyridin-3-yl)trifluoroborate 95%
Potassium (5-bromo-2-fluoropyridin-3-yl)trifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₂BBrF₄KN
MW: 281,88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18325149
CAS Number: 1245906-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR