6-Chloro-8-methoxyquinoline 98%

BLDPBD247121-250MGEA 44.1 EUR
BLDPBD247121-250MG BLDPBD247121-1G BLDPBD247121-100MG
6-Chloro-8-methoxyquinoline 98%
6-Chloro-8-methoxyquinoline

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₈ClNO
MW: 193,63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22200018
CAS Number: 1355066-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR