Ethyl 5-ethyl-1H-imidazole-2-carboxylate 95%

BD247245-1G BD247245-250MG BD247245-100MG
BLDPBD247245-1GEA 743 EUR
BLDPBD247245-1G BLDPBD247245-250MG BLDPBD247245-100MG
Ethyl 5-ethyl-1H-imidazole-2-carboxylate 95%
Ethyl 5-ethyl-1H-imidazole-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₂N₂O₂
MW: 168,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24386911
CAS Number: 1171124-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR