2-(2-Methoxyphenyl)thiazole-4-carbaldehyde 98%

BLDPBD24725-250MGEA 142 EUR
BLDPBD24725-250MG BLDPBD24725-100MG BLDPBD24725-1G
2-(2-Methoxyphenyl)thiazole-4-carbaldehyde 98%
2-(2-Methoxyphenyl)thiazole-4-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₁H₉NO₂S
MW: 219,26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06738365
CAS Number: 885279-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR