(S)-3-Amino-1-methylazepan-2-one hydrochloride 97%

BLDPBD247286-1GEA 267 EUR
BLDPBD247286-1G BLDPBD247286-100MG BLDPBD247286-5G BLDPBD247286-250MG
(S)-3-Amino-1-methylazepan-2-one hydrochloride 97%
(S)-3-Amino-1-methylazepan-2-one hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₁₅ClN₂O
MW: 178,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18909646
CAS Number: 956109-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR