5-Chloro-2-methyl-1H-pyrrolo[2,3-b]pyridine 98%

BLDPBD247877-50MGEA 59.9 EUR
BLDPBD247877-50MG BLDPBD247877-250MG BLDPBD247877-1G BLDPBD247877-100MG
5-Chloro-2-methyl-1H-pyrrolo[2,3-b]pyridine 98%
5-Chloro-2-methyl-1H-pyrrolo[2,3-b]pyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₇ClN₂
MW: 166,60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040729
CAS Number: 145934-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR