(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-(4-iodophenyl)propanoate 98%

BLDPBD247949-100MGEA 67.2 EUR
BLDPBD247949-100MG BLDPBD247949-1G BLDPBD247949-250MG
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-(4-iodophenyl)propanoate 98%
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-(4-iodophenyl)propanoate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₅H₂₀INO₄
MW: 405,22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07366813
CAS Number: 113850-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR