tert-Butyl 2,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride 98%

BLDPBD248571-100MGEA 64.1 EUR
BLDPBD248571-100MG BLDPBD248571-250MG
tert-Butyl 2,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride 98%
tert-Butyl 2,8-diazaspiro[4.5]decane-8-carboxylate hydrochloride
Formula: C₁₃H₂₅ClN₂O₂
MW: 276,8 g/mol
MDL Number: MFCD13185062
CAS Number: 851325-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR