1-Ethyl-1-methyl-4-oxopiperidin-1-ium iodide 95%

BLDPBD248639-250MGEA 50.4 EUR
BLDPBD248639-250MG BLDPBD248639-5G BLDPBD248639-1G
1-Ethyl-1-methyl-4-oxopiperidin-1-ium iodide 95%
1-Ethyl-1-methyl-4-oxopiperidin-1-ium iodide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₁₆INO
MW: 269,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16620306
CAS Number: 77542-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR