4,6-Dichloro-2,3-dimethylpyridine 97%

BLDPBD248830-250MGEA 178 EUR
BLDPBD248830-250MG BLDPBD248830-100MG
4,6-Dichloro-2,3-dimethylpyridine 97%
4,6-Dichloro-2,3-dimethylpyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₇Cl₂N
MW: 176,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26743449
CAS Number: 101252-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR