1-(2-Amino-4-(benzyloxy)-5-methoxyphenyl)ethanone 97%

BD249045-100MG BD249045-250MG
BLDPBD249045-100MGEA 68.3 EUR
BLDPBD249045-100MG BLDPBD249045-250MG
1-(2-Amino-4-(benzyloxy)-5-methoxyphenyl)ethanone 97%
1-(2-Amino-4-(benzyloxy)-5-methoxyphenyl)ethanone

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₆H₁₇NO₃
MW: 271,31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21606560
CAS Number: 75665-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR