(5-Cyanobenzo[b]thiophen-2-yl)boronic acid 95+%

BD249187-250MG BD249187-1G BD249187-50MG BD249187-100MG
BLDPBD249187-250MGEA 303 EUR
BLDPBD249187-250MG BLDPBD249187-1G BLDPBD249187-50MG BLDPBD249187-100MG
(5-Cyanobenzo[b]thiophen-2-yl)boronic acid 95+%
(5-Cyanobenzo[b]thiophen-2-yl)boronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₉H₆BNO₂S
MW: 203,03 g/mol
Storage Temperature: Freezer
MDL Number: MFCD15143421
CAS Number: 1142946-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR