2,2,2,3',5'-Pentafluoroacetophenone 97%

Synonyms: 1-(3,5-Difluorophenyl)-2,2,2-trifluoroethanone

BLDPBD24953-25GEA 162 EUR
BLDPBD24953-25G BLDPBD24953-1G BLDPBD24953-100G BLDPBD24953-5G BLDPBD24953-10G BLDPBD24953-0.025G
2,2,2,3',5'-Pentafluoroacetophenone 97%
2,2,2,3',5'-Pentafluoroacetophenone

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₃F₅O
MW: 210,1000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01319993
CAS Number: 845823-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR