Ethyl 3-Bromopyridine-2-carboxylate 97%

Synonyms: Ethyl 3-bromopicolinate

BLDPBD250575-25GEA 395 EUR
BLDPBD250575-25G BLDPBD250575-250MG BLDPBD250575-5G BLDPBD250575-10G BLDPBD250575-1G
Ethyl 3-Bromopyridine-2-carboxylate 97%
Ethyl 3-Bromopyridine-2-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₈BrNO₂
MW: 230,059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189685
CAS Number: 434319-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR