3,3-Dimethylcyclopentanone 97%

BD251221-250MG BD251221-5G BD251221-1G
BLDPBD251221-250MGEA 26.3 EUR
BLDPBD251221-250MG BLDPBD251221-5G BLDPBD251221-1G
3,3-Dimethylcyclopentanone 97%
3,3-Dimethylcyclopentanone

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₁₂O
MW: 112,16 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18917067
CAS Number: 20500-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR