(R)-5-Bromo-1-methyl-2-tritylisoindoline 95%

BLDPBD251274-100MGEA 55.7 EUR
BLDPBD251274-100MG BLDPBD251274-250MG BLDPBD251274-5G BLDPBD251274-1G
(R)-5-Bromo-1-methyl-2-tritylisoindoline 95%
(R)-5-Bromo-1-methyl-2-tritylisoindoline

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₈H₂₄BrN
MW: 454,40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12974714
CAS Number: 194805-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR