5-Bromo-4-(tert-butyl)pyrimidine 97%

BLDPBD253418-100MGEA 108 EUR
BLDPBD253418-100MG BLDPBD253418-1G BLDPBD253418-5G BLDPBD253418-250MG
5-Bromo-4-(tert-butyl)pyrimidine 97%
5-Bromo-4-(tert-butyl)pyrimidine

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₁BrN₂
MW: 215,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17012062
CAS Number: 1439-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR