cis-Ethyl 3-aminocyclobutanecarboxylate hydrochloride 97%

BD253657-1G BD253657-5G BD253657-100MG BD253657-250MG
BLDPBD253657-1GEA 245 EUR
BLDPBD253657-1G BLDPBD253657-5G BLDPBD253657-100MG BLDPBD253657-250MG
cis-Ethyl 3-aminocyclobutanecarboxylate hydrochloride 97%
cis-Ethyl 3-aminocyclobutanecarboxylate hydrochloride

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₄ClNO₂
MW: 179,65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12925784
CAS Number: 957793-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR