3,5-Dimethoxy-4-methylbenzaldehyde 95%

BLDPBD253943-100MGEA 159 EUR
BLDPBD253943-100MG BLDPBD253943-250MG BLDPBD253943-50MG
3,5-Dimethoxy-4-methylbenzaldehyde 95%
3,5-Dimethoxy-4-methylbenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₀H₁₂O₃
MW: 180,20 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12025247
CAS Number: 1011-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR