4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenol 98%

BD254600-250MG BD254600-1G BD254600-100MG
BLDPBD254600-250MGEA 75.6 EUR
BLDPBD254600-250MG BLDPBD254600-1G BLDPBD254600-100MG
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenol 98%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)phenol

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₃H₁₆BF₃O₃
MW: 288,07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16994250
CAS Number: 1243143-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR